missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-BOC-L-tert-Leucine, 98%, Thermo Scientific Chemicals
35.30 € - 305.00 €
Chemical Identifiers
CAS | 62965-35-9 |
---|---|
Molecular Formula | C11H21NO4 |
Molecular Weight (g/mol) | 231.29 |
MDL Number | MFCD00065574 |
InChI Key | LRFZIPCTFBPFLX-JLDDOWRYNA-N |
Synonym | n-boc-l-tert-leucine, boc-tle-oh, boc-l-tert-leucine, s-n-tert-butoxycarbonyl-tert-leucine, s-2-tert-butoxycarbonyl amino-3,3-dimethylbutanoic acid, boc-l-alpha-t-butylglycine, boc-alpha-t-butylglycine, boc-tbu-gly-oh, boc-l-tle-oh, n-tert-butoxycarbonyl-l-tert-leucine |
PubChem CID | 2734668 |
SMILES | CC(C)(C)OC(=O)N[C@H](C(O)=O)C(C)(C)C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10468172
|
Thermo Scientific Acros
368010010 |
1 g | Glass Bottle |
35.30 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10180762
|
Thermo Scientific Acros
368010050 |
5 g | Glass Bottle |
92.40 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10340634
|
Thermo Scientific Acros
368010250 |
25 g | Glass Bottle |
305.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
62965-35-9 | |
231.29 | |
LRFZIPCTFBPFLX-JLDDOWRYNA-N | |
2734668 |
C11H21NO4 | |
MFCD00065574 | |
n-boc-l-tert-leucine, boc-tle-oh, boc-l-tert-leucine, s-n-tert-butoxycarbonyl-tert-leucine, s-2-tert-butoxycarbonyl amino-3,3-dimethylbutanoic acid, boc-l-alpha-t-butylglycine, boc-alpha-t-butylglycine, boc-tbu-gly-oh, boc-l-tle-oh, n-tert-butoxycarbonyl-l-tert-leucine | |
CC(C)(C)OC(=O)N[C@H](C(O)=O)C(C)(C)C |
Specifications
118.0°C to 121.0°C | |
97.5 | |
White to Yellow | |
98% | |
MFCD00065574 | |
n-boc-l-tert-leucine, boc-tle-oh, boc-l-tert-leucine, s-n-tert-butoxycarbonyl-tert-leucine, s-2-tert-butoxycarbonyl amino-3,3-dimethylbutanoic acid, boc-l-alpha-t-butylglycine, boc-alpha-t-butylglycine, boc-tbu-gly-oh, boc-l-tle-oh, n-tert-butoxycarbonyl-l-tert-leucine | |
CC(C)(C)OC(=O)N[C@H](C(O)=O)C(C)(C)C | |
231.29 | |
231.29 | |
Crystalline Powder |
62965-35-9 | |
100.0 | |
Authentic | |
C11H21NO4 | |
-4.0° to -5.0° (20°C, 589nm) (c=1, acetic acid) | |
LRFZIPCTFBPFLX-JLDDOWRYNA-N | |
(2S)-2-{[(tert-butoxy)carbonyl]amino}-3,3-dimethylbutanoic acid | |
2734668 | |
98% | |
N-BOC-L-tert-Leucine |
Safety and Handling
Product Identifier
- N-BOC-L-tert-Leucine
RUO – Research Use Only