missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Boc-L-threonine methyl ester, 95%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
N-Boc-L-threonine methyl ester is used as pharmaceutical intermediates and chemical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N-Boc-L-threonine methyl ester is used as pharmaceutical intermediates and chemical intermediate.
Solubility
Slightly soluble in water.
Notes
Storage temperature 2 - 4°C. Keep container tightly closed in a dry and well-ventilated place.
N-Boc-L-threonine methyl ester is used as pharmaceutical intermediates and chemical intermediate.
Solubility
Slightly soluble in water.
Notes
Storage temperature 2 - 4°C. Keep container tightly closed in a dry and well-ventilated place.
Specifications
Specifications
Boiling Point | 206°C |
Color | Yellow |
Refractive Index | 1.45 |
Solubility Information | Slightly soluble in water. |
Quantity | 5 g |
Formula Weight | 233.26 |
Percent Purity | 95% |
Physical Form | Viscous Liquid |
Chemical Name or Material | N-Boc-L-threonine methyl ester |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction