missing translation for 'onlineSavingsMsg'
Learn More

N-BOC-L-Valinol, 96%, Thermo Scientific™

Product Code. 10614365
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 79069-14-0
Molecular Formula C10H21NO3
Molecular Weight (g/mol) 203.28
MDL Number MFCD00082635
InChI Key OOQRRYDVICNJGC-MRVPVSSYSA-N
Synonym n-boc-l-valinol, boc-l-valinol, boc-valinol, n-tert-butoxycarbonyl-l-valinol, s-n-tert-butoxycarbonyl valinol, r-n-tert-butoxycarbonyl valinol, tert-butyl n-2s-1-hydroxy-3-methylbutan-2-yl carbamate, boc-d-val-ol, boc-valch2oh, n-t-boc-l-valinol
PubChem CID 7021464
IUPAC Name tert-butyl N-[(2S)-1-hydroxy-3-methylbutan-2-yl]carbamate
SMILES CC(C)[C@@H](CO)NC(=O)OC(C)(C)C

Specifications

Packaging Glass bottle
CAS Min % 95.0
CAS Max % 100.0
Density 0.9950g/mL
Boiling Point 218.0°C
Quantity 5 g
Flash Point 85°C
Assay Percent Range 96%
Formula Weight 203.28
Percent Purity 96%
Chemical Name or Material N-BOC-L-Valinol
Show More Show Less
compliance-icons
Product Identifier
  • N-BOC-L-Valinol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.