missing translation for 'onlineSavingsMsg'
Learn More
N-Boc-N-methyl-L-valine, 95%, Thermo Scientific Chemicals Product Code.: 15499248

N-Boc-N-methyl-L-valine, 95%, Thermo Scientific Chemicals

Product Code. 15499248
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 15499248

Brand: Thermo Scientific Alfa Aesar H62771.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 45170-31-8
Molecular Formula C11H21NO4
Molecular Weight (g/mol) 231.29
MDL Number MFCD00038760
InChI Key XPUAXAVJMJDPDH-QMMMGPOBSA-N
Synonym boc-n-me-val-oh, boc-n-methyl-l-valine, n-boc-n-methyl-l-valine, boc-meval-oh, s-2-tert-butoxycarbonyl methyl amino-3-methylbutanoic acid, l-valine, n-1,1-dimethylethoxy carbonyl-n-methyl, 2s-2-tert-butoxycarbonyl methyl amino-3-methylbutanoic acid, ambotzbaa1272, n-boc-n-methylvaline, pubchem12254
PubChem CID 7010608
IUPAC Name (2S)-3-methyl-2-[methyl-[(2-methylpropan-2-yl)oxycarbonyl]amino]butanoic acid
SMILES CC(C)[C@H](N(C)C(=O)OC(C)(C)C)C(O)=O

Specifications

Beilstein 2646180
Quantity 1 g
Formula Weight 231.29
Percent Purity 95%
Physical Form Solid
Chemical Name or Material N-Boc-N-methyl-L-valine

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberR12I017 Certificate TypeCertificate of Analysis Date11/08/2022 Catalog NumberH62771.06,H62771.03
Lot NumberX11D044 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH62771.06,H62771.03
Lot NumberT02H026 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH62771.06,H62771.03
Lot NumberQ27G019 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH62771.06,H62771.03

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
N-Boc-N-methyl-L-valine, 95%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.