missing translation for 'onlineSavingsMsg'
Learn More
N-Boc-O-benzyl-D-tyrosine, 95%, Thermo Scientific Chemicals Product Code.: 15450119

N-Boc-O-benzyl-D-tyrosine, 95%, Thermo Scientific Chemicals

Product Code. 15450119
25 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 15450119

Brand: Thermo Scientific Alfa Aesar H63427.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

N-Boc-O-benzyl-D-tyrosine is used as a intermediate for pharmaceutical and chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-O-benzyl-D-tyrosine is used as a intermediate for pharmaceutical and chemical research.

Solubility
Slightly soluble in water.

Notes
Store in freezer at(-20°C). Keep container tightly closed. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63769-58-4
Molecular Formula C21H25NO5
Molecular Weight (g/mol) 371.43
MDL Number MFCD00038249
InChI Key ZAVSPTOJKOFMTA-LGWFVXIRNA-N
Synonym boc-d-tyr bzl-oh, boc-o-benzyl-d-tyrosine, n-boc-o-benzyl-d-tyrosine, boc-o-benzyl-d-tyr, r-3-4-benzyloxy phenyl-2-tert-butoxycarbonyl amino propanoic acid, 2r-3-4-benzyloxy phenyl-2-tert-butoxycarbonyl amino propanoic acid, ambotzbaa1381, boc-dtyr bn-oh, pubchem14949, boc-d-tyr bzl oh
PubChem CID 6992575
IUPAC Name (2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-(4-phenylmethoxyphenyl)propanoic acid
SMILES CC(C)(C)OC(=O)N[C@@H](CC1=CC=C(OCC2=CC=CC=C2)C=C1)C(O)=O

Specifications

Solubility Information Slightly soluble in water.
Quantity 25 g
Formula Weight 371.43
Percent Purity 95%
Physical Form Solid
Chemical Name or Material N-Boc-O-benzyl-D-tyrosine

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot Number81500085 Certificate TypeCertificate of Analysis Date18/01/2025 Catalog NumberH63427.06,H63427.14
Lot Number81500159 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberH63427.06,H63427.14
Lot Number81500085 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberH63427.06,H63427.14
Lot Number81301468 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberH63427.06,H63427.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
N-Boc-O-benzyl-D-tyrosine, 95%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.