missing translation for 'onlineSavingsMsg'
Learn More
N-Boc-O-benzyl-L-serine, 97%, Thermo Scientific Chemicals Product Code.: 15449188

N-Boc-O-benzyl-L-serine, 97%, Thermo Scientific Chemicals

Product Code. 15449188
25 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 15449188

Brand: Thermo Scientific Alfa Aesar H62705.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23680-31-1
Molecular Formula C15H21NO5
Molecular Weight (g/mol) 295.335
MDL Number MFCD00066063
InChI Key DMBKPDOAQVGTST-LBPRGKRZSA-N
Synonym boc-ser bzl-oh, n-boc-o-benzyl-l-serine, boc-o-benzyl-l-serine, boc-ser bzl, s-3-benzyloxy-2-tert-butoxycarbonyl amino propanoic acid, n-1,1-dimethylethoxy carbonyl-o-phenylmethyl-l-serine, n-t-butoxycarbonyl-o-benzyl-l-serine, o-benzyl-n-tert-butyloxycarbonylserine, n-tert-butoxycarbonyl-o-benzyl-l-serine, o-benzyl-n-tert-butoxycarbonyl-l-serine
PubChem CID 90234
IUPAC Name (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-phenylmethoxypropanoic acid
SMILES CC(C)(C)OC(=O)NC(COCC1=CC=CC=C1)C(=O)O

Specifications

Melting Point 58°C to 60°C
Beilstein 3064461
Quantity 25 g
Formula Weight 295.33
Percent Purity 97%
Physical Form Solid
Chemical Name or Material N-Boc-O-benzyl-L-serine

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot Number5007I22X Certificate TypeCertificate of Analysis Date04/10/2023 Catalog NumberH62705.06,H62705.14,H62705.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
N-Boc-O-benzyl-L-serine, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.