missing translation for 'onlineSavingsMsg'
Learn More
Learn More
n-Butanol, 99% max., Molecular biology reagent grade, MP Biomedicals™
Click to view available options
Quantity:
1 L
Specifications
Specifications
Density | 0.81g/cm3 at 21°C |
Quantity | 1 L |
Physical Form | Liquid |
Formula Weight | 74.12 |
Percent Purity | 99% max. |
Grade | Molecular Biology Reagent |
Chemical Name or Material | n-Butanol |
Safety and Handling
Signal Word
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H226-Flammable liquid and vapour.
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H335-May cause respiratory irritation.
- H336-May cause drowsiness or dizziness.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
n-Butanol, 99% max., Molecular biology reagent grade, MP Biomedicals™ >
Spot an opportunity for improvement?Share a Content Correction