Learn More
n-Butyl acetate, HPLC Grade, 99.5+%, Thermo Scientific Chemicals
47.34 € - 254.27 €
Chemical Identifiers
CAS | 123-86-4 |
---|---|
Molecular Formula | C6H12O2 |
Molecular Weight (g/mol) | 116.16 |
MDL Number | MFCD00009445 |
InChI Key | DKPFZGUDAPQIHT-UHFFFAOYSA-N |
Synonym | n-butyl acetate, acetic acid, butyl ester, 1-butyl acetate, butyl ethanoate, acetic acid butyl ester, n-butyl ethanoate, n-butylacetate, butylacetat, acetic acid n-butyl ester, acetate de butyle |
PubChem CID | 31272 |
ChEBI | CHEBI:31328 |
IUPAC Name | butyl acetate |
SMILES | CCCCOC(C)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11313588
|
Thermo Scientific Alfa Aesar
039197.AK |
250 mL |
47.34 €
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11383578
|
Thermo Scientific Alfa Aesar
039197.K2 |
1 L |
109.55 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11393578
|
Thermo Scientific Alfa Aesar
039197.M1 |
4 x 1 L |
254.27 €
4 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
n-Butyl acetate is used as a mobile phase in High Performance Liquid Chromatography and Liquid Chromatography coupled with Mass Spectrometry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
123-86-4 | |
116.16 | |
DKPFZGUDAPQIHT-UHFFFAOYSA-N | |
31272 | |
butyl acetate |
C6H12O2 | |
MFCD00009445 | |
n-butyl acetate, acetic acid, butyl ester, 1-butyl acetate, butyl ethanoate, acetic acid butyl ester, n-butyl ethanoate, n-butylacetate, butylacetat, acetic acid n-butyl ester, acetate de butyle | |
CHEBI:31328 | |
CCCCOC(C)=O |
Specifications
123-86-4 | |
Colorless | |
5 | |
25°C (77°F) | |
C6H12O2 | |
1.3945 | |
UN1123 | |
14,1535 | |
Miscible with ethanol,ethyl ether,alcohol,acetone,chloroform and most hydrocarbonsSlightly miscible with water. | |
CCCCOC(C)=O | |
116.16 | |
CHEBI:31328 | |
≥99.5% | |
Liquid |
-78°C | |
0.881 g/mL | |
124°C to 126°C | |
Fruit-like | |
CH3CO2(CH2)3CH3 | |
MFCD00009445 | |
1741921 | |
n-butyl acetate, acetic acid, butyl ester, 1-butyl acetate, butyl ethanoate, acetic acid butyl ester, n-butyl ethanoate, n-butylacetate, butylacetat, acetic acid n-butyl ester, acetate de butyle | |
DKPFZGUDAPQIHT-UHFFFAOYSA-N | |
butyl acetate | |
31272 | |
116.16 | |
HPLC | |
n-Butyl acetate |
Safety and Handling
GHS H Statement
H226-H336
Flammable liquid and vapor.
May cause drowsiness or dizziness.
P210-P233-P235-P240-P241-P242-P243-P261-P271-P280-P303+P361+P353-P304+P340-P312-P370+P378q-P501c
H226-H336
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: BUTYL ACETATES
missing translation for 'einecsNumber' : 204-658-1
missing translation for 'rtecsNumber' : AF350000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- n-Butyl acetate, HPLC Grade
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
- Specific target organ toxicity Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
- H336-May cause drowsiness or dizziness.
- EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
Supplemental information
- MIXTURE LIST-Contains : n-Butyl acetate
RUO – Research Use Only