missing translation for 'onlineSavingsMsg'
Learn More

n-Butyl disulfide, 97%, Thermo Scientific Chemicals

Product Code. 10539820
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 629-45-8
Molecular Formula C8H18S2
Molecular Weight (g/mol) 178.35
MDL Number MFCD00009467
InChI Key CUDSBWGCGSUXDB-UHFFFAOYSA-N
Synonym dibutyl disulfide, butyl disulfide, disulfide, dibutyl, n-butyl disulfide, di-n-butyl disulfide, dibutyl disulphide, 5,6-dithiadecane, 1-butyldisulfanyl butane, unii-z77ef35c0q, 1-butyldisulfanyl-butane
PubChem CID 12386
IUPAC Name 1-(butyldisulfanyl)butane
SMILES CCCCSSCCCC

Specifications

Color Brown to Colorless
Density 0.9380g/mL
Boiling Point 229.0°C to 233.0°C
Flash Point 93°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula [CH3(CH2)3]2S2
Refractive Index 1.4915 to 1.4935
Quantity 25 g
Beilstein 01,II,4
Specific Gravity 0.938
Solubility Information Solubility in water: immiscible
Formula Weight 178.35
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material n-Butyl disulfide
Show More Show Less
compliance-icons
Product Identifier
  • n-Butyl disulfide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.