missing translation for 'onlineSavingsMsg'
Learn More
n-Butyldimethylchlorosilane, 96%, Thermo Scientific Chemicals Product Code.: 11405577

n-Butyldimethylchlorosilane, 96%, Thermo Scientific Chemicals

Product Code. 11405577
25 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11405577

Brand: Thermo Scientific Alfa Aesar B23551.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1000-50-6
Molecular Formula C6H15ClSi
Molecular Weight (g/mol) 150.72
MDL Number MFCD00015727
InChI Key MXOSTENCGSDMRE-UHFFFAOYSA-N
Synonym butylchlorodimethylsilane, butyldimethylchlorosilane, n-butyldimethylchlorosilane, butyldimethylsilyl chloride, 1-butyldimethylchlorosilane, silane, butylchlorodimethyl, butyl chloro dimethylsilane, n-butyl-dimethylchlorosilane, butyl-chloro-dimethyl-silane, n-butyldimethylsilyl chloride
PubChem CID 66079
SMILES CCCC[Si](C)(C)Cl

Specifications

Boiling Point 138°C
Quantity 25 g
UN Number UN2920
Beilstein 1732573
Flash Point 27°C (80°F)
IUPAC Name butyl-chloro-dimethylsilane
Formula Weight 150.73
Percent Purity 96%
Refractive Index 1.42
Sensitivity Moisture sensitive
Density 0.875
Chemical Name or Material n-Butyldimethylchlorosilane
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0294858 Certificate TypeCertificate of Analysis Date09/10/2023 Catalog NumberB23551.06,B23551.14,B23551.22
Lot NumberG6511A Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB23551.06,B23551.14,B23551.22
Lot Number10161726 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB23551.06,B23551.14,B23551.22
Lot Number10187528 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB23551.06,B23551.14,B23551.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
n-Butyldimethylchlorosilane, 96%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.