Learn More
n-Butylmagnesium chloride, 20 wt% solution in THF/Toluene, AcroSeal™, Thermo Scientific Chemicals
67.70 € - 279.00 €
Chemical Identifiers
CAS | 693-04-9, 109-99-9, 108-88-3 |
---|---|
Molecular Formula | C4H9ClMg |
Molecular Weight (g/mol) | 116.87 |
MDL Number | MFCD00000475 |
InChI Key | UUNGXYVLXYGZKH-UHFFFAOYSA-M |
Synonym | butylmagnesium chloride, magnesium, butylchloro, nbumgcl, bumgcl, butylmagnesiumchloride, n-butylmagnesium chloride, n-butyl-magnesium chloride, butylmagnesium chloride solution, butylmagnesium chloride solution, 2.0 m in diethyl ether, unii-4j83fgw852 |
PubChem CID | 101837 |
SMILES | CCCC[Mg]Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10700611
|
Thermo Scientific Acros
331681000 |
100 mL | AcroSeal™ Glass bottle |
67.70 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11915851
|
Thermo Scientific Acros
331688000 |
800 mL | AcroSeal™ Glass Bottle |
279.00 €
800mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
693-04-9, 109-99-9, 108-88-3 | |
116.87 | |
UUNGXYVLXYGZKH-UHFFFAOYSA-M | |
101837 |
C4H9ClMg | |
MFCD00000475 | |
butylmagnesium chloride, magnesium, butylchloro, nbumgcl, bumgcl, butylmagnesiumchloride, n-butylmagnesium chloride, n-butyl-magnesium chloride, butylmagnesium chloride solution, butylmagnesium chloride solution, 2.0 m in diethyl ether, unii-4j83fgw852 | |
CCCC[Mg]Cl |
Specifications
693-04-9, 109-99-9, 108-88-3 | |
19.0, 78.0, 78.0 | |
Solution | |
CH3(CH2)3MgCl | |
−21°C | |
UUNGXYVLXYGZKH-UHFFFAOYSA-M | |
magnesium;butane;chloride | |
101837 | |
19 to 22% (as BUMC) | |
1.0000g/mL |
Brown to Brown-Green | |
22.0, 81.0, 81.0 | |
C4H9ClMg | |
MFCD00000475 | |
butylmagnesium chloride, magnesium, butylchloro, nbumgcl, bumgcl, butylmagnesiumchloride, n-butylmagnesium chloride, n-butyl-magnesium chloride, butylmagnesium chloride solution, butylmagnesium chloride solution, 2.0 m in diethyl ether, unii-4j83fgw852 | |
CCCC[Mg]Cl | |
116.87 | |
116.87 | |
1 | |
n-Butylmagnesium chloride |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
May be fatal if swallowed and enters airways.
May cause drowsiness or dizziness.
Suspected of damaging the unborn child.
May cause damage to organs through prolonged
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Handle under inert gas.
Protect from moisture
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 211-739-5
Recommended Storage : Product may develop some turbidity or precipitate
- 3,5-Dimethylphenol
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- H311-Toxic in contact with skin.
- H301-Toxic if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
RUO – Research Use Only