missing translation for 'onlineSavingsMsg'
Learn More

n-Butyltin hydroxide oxide, 95%, Thermo Scientific Chemicals

Product Code. 11443837
Click to view available options
Quantity:
100 g
500 g
2500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2273-43-0
Molecular Formula C4H10O2Sn
Molecular Weight (g/mol) 208.83
MDL Number MFCD00013927
InChI Key WIHMDCQAEONXND-UHFFFAOYSA-M
Synonym stannane, butylhydroxyoxo, butyltin hydroxide oxide, butylstannoic acid, butylstannonic acid, 1-butanestannonic acid, n-butyltin hydroxide oxide, butylhydroxytin oxide, butylhydroxyoxostannane, unii-g34wda7z2e, butylenestannonic acid
PubChem CID 16767
SMILES CCCC[Sn](O)=O

Specifications

Quantity 100 g
UN Number UN3146
Sensitivity Hygroscopic
IUPAC Name butyl(hydroxy)stannanone
Formula Weight 208.81
Percent Purity 95%
Chemical Name or Material n-Butyltin hydroxide oxide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'