missing translation for 'onlineSavingsMsg'
Learn More

n-Butyltin trichloride, 96%, Thermo Scientific Chemicals

Product Code. 11314819
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1118-46-3
Molecular Formula C4H9Cl3Sn
Molecular Weight (g/mol) 282.176
MDL Number MFCD00000515
InChI Key YMLFYGFCXGNERH-UHFFFAOYSA-K
Synonym butyltin trichloride, stannane, butyltrichloro, monobutyltin trichloride, butyltrichlorotin, trichlorobutyltin, butyl trichloro stannane, n-butyltin trichloride, trichlorobutylstannane, stannane, trichlorobutyl, monotributyltin trichloride
PubChem CID 14236
IUPAC Name butyl(trichloro)stannane
SMILES CCCC[Sn](Cl)(Cl)Cl

Specifications

Melting Point -63°C
Density 1.693 g/mL
Boiling Point 98°C (10 mmHg)
Flash Point 81°C (178°F)
Odor Acrid
Refractive Index 1.5244
Quantity 25 g
UN Number UN3265
Sensitivity Moisture sensitive
Formula Weight 282.17
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material n-Butyltin trichloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'