missing translation for 'onlineSavingsMsg'
Learn More

N-Chlorosuccinimide, 97%

Product Code. 10396640 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
100g
2.5kg
500g
This item is not returnable. View return policy

Product Code. 10396640

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 128-09-6 | C4H4ClNO2 | 133.53 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 128-09-6
Molecular Formula C4H4ClNO2
Molecular Weight (g/mol) 133.53
InChI Key JRNVZBWKYDBUCA-UHFFFAOYSA-N
Synonym n-chlorosuccinimide, succinchlorimide, chlorosuccinimide, succinochlorimide, succinic n-chloroimide, 2,5-pyrrolidinedione, 1-chloro, 1-chloro-2,5-pyrrolidinedione, succinimide, n-chloro, ncs, caswell no. 807
PubChem CID 31398
ChEBI CHEBI:53203
IUPAC Name 1-chloropyrrolidine-2,5-dione
SMILES C1CC(=O)N(C1=O)Cl

Specifications

Melting Point 144.0°C to 150.0°C
Color White
Flash Point >110°C
Loss on Drying 4% (2g, 110°C, 2 hrs) max.
Infrared Spectrum Authentic
Assay Percent Range 96% min. (on dry substance) (Iodometry)
Packaging Plastic bottle
Quantity 100 g
Beilstein 21,38
Fieser 01,139; 02,69; 05,127; 06,115; 07,65; 08,103; 09,111; 10,94; 12,121; 13,62; 14,87; 15,86; 16,84
Merck Index 15,2167
Solubility Information Solubility in water: 14g/L (20°C). Other solubilities: sparingly soluble in chloroform,ether and ccl4,1g/150mL alcohol- 1g/50mL benzene,soluble in acetone
Formula Weight 133.53
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material N-Chlorosuccinimide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • N-Chlorosuccinimide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.