Learn More
n-Decyldimethylchlorosilane, 97%, Thermo Scientific™
79.00 € - 255.00 €
Chemical Identifiers
CAS | 38051-57-9 |
---|---|
Molecular Formula | C12H27ClSi |
Molecular Weight (g/mol) | 234.883 |
MDL Number | MFCD00054168 |
InChI Key | ZLZGHBNDPINFKG-UHFFFAOYSA-N |
Synonym | chloro decyl dimethylsilane, n-decyldimethylchlorosilane, decyldimethylchlorosilane, dimethyldecylchlorosilane, acmc-209ixd, 1-chlorodimethylsilyldecane, decyl-dimethyl-chlorosilane, 1-chlorodimethylsilyl decane, 10-chlorodecyl-dimethylsilicon, zlzghbndpinfkg-uhfffaoysa |
PubChem CID | 2758020 |
IUPAC Name | chloro-decyl-dimethylsilane |
SMILES | CCCCCCCCCC[Si](C)(C)Cl |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
38051-57-9 | |
234.883 | |
ZLZGHBNDPINFKG-UHFFFAOYSA-N | |
2758020 | |
CCCCCCCCCC[Si](C)(C)Cl |
C12H27ClSi | |
MFCD00054168 | |
chloro decyl dimethylsilane, n-decyldimethylchlorosilane, decyldimethylchlorosilane, dimethyldecylchlorosilane, acmc-209ixd, 1-chlorodimethylsilyldecane, decyl-dimethyl-chlorosilane, 1-chlorodimethylsilyl decane, 10-chlorodecyl-dimethylsilicon, zlzghbndpinfkg-uhfffaoysa | |
chloro-decyl-dimethylsilane |
Specifications
98°C (2mmHg) | |
Liquid | |
MFCD00054168 | |
137°C (279°F) | |
ZLZGHBNDPINFKG-UHFFFAOYSA-N | |
chloro-decyl-dimethylsilane | |
2758020 | |
97% | |
Moisture Sensitive | |
n-Decyldimethylchlorosilane |
38051-57-9 | |
C12H27ClSi | |
UN1760 | |
chloro decyl dimethylsilane, n-decyldimethylchlorosilane, decyldimethylchlorosilane, dimethyldecylchlorosilane, acmc-209ixd, 1-chlorodimethylsilyldecane, decyl-dimethyl-chlorosilane, 1-chlorodimethylsilyl decane, 10-chlorodecyl-dimethylsilicon, zlzghbndpinfkg-uhfffaoysa | |
CCCCCCCCCC[Si](C)(C)Cl | |
234.883 | |
234.88 | |
1.441 | |
0.866g/mL |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, N.O.S.
missing translation for 'tsca' : TSCA
Your input is important to us. Please complete this form to provide feedback related to the content on this product.