missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals N(epsilon)-Boc-L-lysine, 97% Product Code.: 11430347

Thermo Scientific Chemicals N(epsilon)-Boc-L-lysine, 97%

Product Code. 11430347
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11430347

Brand: Thermo Scientific Chemicals B21738.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

It is used as therapy to treat recurrent herpes simplex infections.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as therapy to treat recurrent herpes simplex infections.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2418-95-3
Molecular Formula C11H22N2O4
Molecular Weight (g/mol) 246.31
MDL Number MFCD00037221
InChI Key VVQIIIAZJXTLRE-QMMMGPOBSA-N
Synonym h-lys boc-oh, lys boc, nepsilon-tert-butoxycarbonyl-l-lysine, n6-boc-l-lysine, n∼6∼-tert-butoxycarbonyl-l-lysine, nepsilon-boc-l-lysine, ne-boc-l-lysine, n epsilon-boc-l-lysine, h-lys boc-2-chlorotrityl resin, n6-tert-butoxycarbonyl-l-lysine
PubChem CID 2733283
IUPAC Name (2S)-2-amino-6-[(2-methylpropan-2-yl)oxycarbonylamino]hexanoic acid
SMILES CC(C)(C)OC(=O)NCCCC[C@H]([NH3+])C([O-])=O

Specifications

Melting Point ∼250°C (decomposition)
Beilstein 4252546
Quantity 1 g
Formula Weight 246.31
Percent Purity 97%
Chemical Name or Material N(epsilon)-Boc-L-lysine

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10243927 Certificate TypeCertificate of Analysis Date20/10/2022 Catalog NumberB21738.03,B21738.06
Lot NumberG6501A Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB21738.03,B21738.06
Lot Number10183669 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB21738.03,B21738.06
Lot Number10158463 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB21738.03,B21738.06
Lot Number10139698 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB21738.03,B21738.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals N(epsilon)-Boc-L-lysine, 97% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.