Learn More
N-Ethylaniline, 97%, Thermo Scientific Chemicals
39.10 € - 106.00 €
Chemical Identifiers
CAS | 103-69-5 |
---|---|
Molecular Formula | C8H11N |
Molecular Weight (g/mol) | 121.183 |
MDL Number | MFCD00009025 |
InChI Key | OJGMBLNIHDZDGS-UHFFFAOYSA-N |
Synonym | ethylaniline, n-ethyl aniline, benzenamine, n-ethyl, anilinoethane, n-ethylbenzenamine, ethylphenylamine, n-ethyl-n-phenylamine, aniline, n-ethyl, n-ethylaminobenzene, aethylanilin |
PubChem CID | 7670 |
ChEBI | CHEBI:34870 |
IUPAC Name | N-ethylaniline |
SMILES | CCNC1=CC=CC=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11467337
|
Thermo Scientific Alfa Aesar
A15060.AP |
500 mL |
39.10 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11477337
|
Thermo Scientific Alfa Aesar
A15060.0F |
2500 mL |
106.00 €
2500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
N-Ethylaniline (NEA) may be used as an internal standard in the GC analysis of nicotine extraction from nicotine gum. It may be used in the synthesis of the following, 2-(N-ethylphenylamino)-1,4-benzoquinone, 2-(arylaminomethyl)phenylboronic acid, poly(methyl methacrylate) (PMMA) films containing NEA and poly(N-ethylaniline) (PNEA).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsN-Ethylaniline (NEA) may be used as an internal standard in the GC analysis of nicotine extraction from nicotine gum. It may be used in the synthesis of the following, 2-(N-ethylphenylamino)-1,4-benzoquinone, 2-(arylaminomethyl)phenylboronic acid, poly(methyl methacrylate) (PMMA) films containing NEA and poly(N-ethylaniline) (PNEA).
Solubility
Soluble in water (50g/L).
Notes
Air and Light Sensitive, store in dark and away from air. Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
Chemical Identifiers
103-69-5 | |
121.183 | |
OJGMBLNIHDZDGS-UHFFFAOYSA-N | |
7670 | |
N-ethylaniline |
C8H11N | |
MFCD00009025 | |
ethylaniline, n-ethyl aniline, benzenamine, n-ethyl, anilinoethane, n-ethylbenzenamine, ethylphenylamine, n-ethyl-n-phenylamine, aniline, n-ethyl, n-ethylaminobenzene, aethylanilin | |
CHEBI:34870 | |
CCNC1=CC=CC=C1 |
Specifications
103-69-5 | |
0.961 | |
85°C (185°F) | |
C8H11N | |
MFCD00009025 | |
507468 | |
14,3764 | |
Soluble in water (50g/L). | |
CCNC1=CC=CC=C1 | |
121.183 | |
CHEBI:34870 | |
97% |
-64°C | |
204°C to 205°C | |
Odorless | |
1.554 | |
UN2272 | |
Air and light sensitive | |
ethylaniline, n-ethyl aniline, benzenamine, n-ethyl, anilinoethane, n-ethylbenzenamine, ethylphenylamine, n-ethyl-n-phenylamine, aniline, n-ethyl, n-ethylaminobenzene, aethylanilin | |
OJGMBLNIHDZDGS-UHFFFAOYSA-N | |
N-ethylaniline | |
7670 | |
121.18 | |
N-Ethylaniline |
Safety and Handling
GHS H Statement
H301-H311-H331-H373-H227
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Combustible liquid.
P210-P235-P260-P264b-P270-P271-P280-P284-P301+P310-P302+P352-P304+P340-P310-P312-P314-P330-P361-P363-P370+P378q-P501c
H227-H301+H311-H330-H373
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: N-ETHYLANILINE
missing translation for 'einecsNumber' : 203-135-5
missing translation for 'rtecsNumber' : BX9780000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only