missing translation for 'onlineSavingsMsg'
Learn More

N-Ethylaniline, 98%, Thermo Scientific Chemicals

Product Code. 10327130
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103-69-5
Molecular Formula C8H11N
Molecular Weight (g/mol) 121.18
MDL Number MFCD00009025
InChI Key OJGMBLNIHDZDGS-UHFFFAOYSA-N
Synonym ethylaniline, n-ethyl aniline, benzenamine, n-ethyl, anilinoethane, n-ethylbenzenamine, ethylphenylamine, n-ethyl-n-phenylamine, aniline, n-ethyl, n-ethylaminobenzene, aethylanilin
PubChem CID 7670
ChEBI CHEBI:34870
IUPAC Name N-ethylaniline
SMILES CCNC1=CC=CC=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -63°C
Color Brown to Yellow
Density 0.9630g/mL
Boiling Point 205°C
Flash Point 85°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H5NHC2H5
Refractive Index 1.5530 to 1.555
Quantity 100 g
Beilstein 12, 159
Merck Index 15, 3819
Specific Gravity 0.963
Solubility Information Solubility in water: 50g/L water (20°C). Other solubilities: soluble in alcohol,ether and many other organic,solvents
Formula Weight 121.18
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material N-Ethylaniline, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • N-Ethylaniline
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H373-May cause damage to organs.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'