missing translation for 'onlineSavingsMsg'
Learn More

N-Ethylcyclohexylamine, 98%, Thermo Scientific™

18.90 € - 43.40 €

Chemical Identifiers

CAS 5459-93-8
Molecular Formula C8H17N
Molecular Weight (g/mol) 127.23
InChI Key AGVKXDPPPSLISR-UHFFFAOYSA-N
Synonym n-ethylcyclohexylamine, cyclohexanamine, n-ethyl, n-cyclohexylethylamine, vulkacit hx, accelerator hx, cyclohexylamine, n-ethyl, ethylamino cyclohexane, cyclohexyl ethyl amine, n-ethyl cyclohexyl amine, unii-yjk13p0h3e
PubChem CID 21609
IUPAC Name N-ethylcyclohexanamine
SMILES CCNC1CCCCC1
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
11386365
View Documents
Thermo Scientific Acros
437740250
25 mL Glass bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11396365
View Documents
Thermo Scientific Acros
437741000
100 mL Glass bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Specifications

Chemical Identifiers

5459-93-8
127.23
n-ethylcyclohexylamine, cyclohexanamine, n-ethyl, n-cyclohexylethylamine, vulkacit hx, accelerator hx, cyclohexylamine, n-ethyl, ethylamino cyclohexane, cyclohexyl ethyl amine, n-ethyl cyclohexyl amine, unii-yjk13p0h3e
N-ethylcyclohexanamine
C8H17N
AGVKXDPPPSLISR-UHFFFAOYSA-N
21609
CCNC1CCCCC1

Specifications

5459-93-8
100.0
0.8500g/mL
30°C
97.5% min. (GC)
1.4500 to 1.4530 (20°C, 589nm)
n-ethylcyclohexylamine, cyclohexanamine, n-ethyl, n-cyclohexylethylamine, vulkacit hx, accelerator hx, cyclohexylamine, n-ethyl, ethylamino cyclohexane, cyclohexyl ethyl amine, n-ethyl cyclohexyl amine, unii-yjk13p0h3e
CCNC1CCCCC1
127.23
127.23
Liquid
97.5
Colorless to Yellow
164°C
Authentic
C8H17N
0.85
AGVKXDPPPSLISR-UHFFFAOYSA-N
N-ethylcyclohexanamine
21609
98%
N-Ethylcyclohexylamine
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes severe skin burns and eye damage.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 226-733-8

Product Identifier
  • N-Ethylcyclohexylamine

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 A

Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.