missing translation for 'onlineSavingsMsg'
Learn More

N-Ethylmethylamine, 97%, Thermo Scientific Chemicals

Product Code. 10115570
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 624-78-2
Molecular Formula C3H9N
Molecular Weight (g/mol) 59.11
MDL Number MFCD00009030
InChI Key LIWAQLJGPBVORC-UHFFFAOYSA-N
Synonym n-ethylmethylamine, methylethylamine, ethanamine, n-methyl, n-methylethylamine, ethyl methyl amine, ethylmethylamine, nmea, n-methyl-n-ethylamine, ethylamine, n-methyl, ethyl-methyl-amine
PubChem CID 12219
IUPAC Name N-methylethanamine
SMILES CCNC

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 0.6800g/mL
Boiling Point 33.0°C to 34.0°C
Flash Point <−34°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula C2H5NHCH3
Refractive Index 1.3730 to 1.3770
Quantity 5 g
Specific Gravity 0.68
Solubility Information Solubility in water: soluble
Formula Weight 59.11
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material N-Ethylmethylamine
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.