missing translation for 'onlineSavingsMsg'
Learn More
N-Ethylmethylamine, 98+%, Thermo Scientific Chemicals Product Code.: 11386916

N-Ethylmethylamine, 98+%, Thermo Scientific Chemicals

Product Code. 11386916
25 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 11386916

Brand: Thermo Scientific Alfa Aesar L04372.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 624-78-2
Molecular Formula C3H9N
Molecular Weight (g/mol) 59.11
MDL Number MFCD00009030
InChI Key LIWAQLJGPBVORC-UHFFFAOYSA-N
Synonym n-ethylmethylamine, methylethylamine, ethanamine, n-methyl, n-methylethylamine, ethyl methyl amine, ethylmethylamine, nmea, n-methyl-n-ethylamine, ethylamine, n-methyl, ethyl-methyl-amineShow MoreShow Less
PubChem CID 12219
IUPAC Name N-methylethanamine
SMILES CCNC

Specifications

Density 0.688
Boiling Point 33°C to 34°C
Flash Point <−34°C (−29°F)
Odor Amine-like
Refractive Index 1.374
Quantity 25 g
UN Number UN2733
Beilstein 1730786
Sensitivity Air sensitive; Hygroscopic
Formula Weight 59.11
Percent Purity ≥98%
Chemical Name or Material N-Ethylmethylamine
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10241195 Certificate TypeCertificate of Analysis Date02/04/2024 Catalog NumberL04372.14,L04372.06
Lot Number10227698 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL04372.14,L04372.06
Lot Number10236581 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL04372.14,L04372.06
Lot Number10193350 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL04372.14,L04372.06
Lot Number10217816 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL04372.14,L04372.06
compliance-icons compliance-icons compliance-icons
Product Identifier
  • N-Ethylmethylamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : N-Ethylmethylamine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
N-Ethylmethylamine, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.