Learn More
N-Ethylmorpholine, 99%, Thermo Scientific Chemicals
21.31 € - 229.14 €
Chemical Identifiers
CAS | 100-74-3 |
---|---|
Molecular Formula | C6H13NO |
Molecular Weight (g/mol) | 115.17 |
MDL Number | MFCD00006177 |
InChI Key | HVCNXQOWACZAFN-UHFFFAOYSA-N |
Synonym | n-ethylmorpholine, morpholine, 4-ethyl, ethylmorpholine, n-ethylmorfolin, n-ethyl morpholine, n-ethylmorfolin czech, unii-ecm0g991fq, n-ethyl-morpholine, ccris 4818, ecm0g991fq |
PubChem CID | 7525 |
IUPAC Name | 4-ethylmorpholine |
SMILES | CCN1CCOCC1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10274810
|
Thermo Scientific Acros
149401000 |
100 mL | Glass bottle |
21.31 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10745041
|
Thermo Scientific Acros
149405000 |
500 mL | Glass bottle |
59.50 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10227213
|
Thermo Scientific Acros
149400025 |
2.5 L | Glass bottle |
229.14 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
100-74-3 | |
115.17 | |
HVCNXQOWACZAFN-UHFFFAOYSA-N | |
7525 | |
CCN1CCOCC1 |
C6H13NO | |
MFCD00006177 | |
n-ethylmorpholine, morpholine, 4-ethyl, ethylmorpholine, n-ethylmorfolin, n-ethyl morpholine, n-ethylmorfolin czech, unii-ecm0g991fq, n-ethyl-morpholine, ccris 4818, ecm0g991fq | |
4-ethylmorpholine |
Specifications
100-74-3 | |
100.0 | |
Brown-Gray to Cream | |
139.0°C | |
Authentic | |
C6H13NO | |
MFCD00006177 | |
01,383 | |
n-ethylmorpholine, morpholine, 4-ethyl, ethylmorpholine, n-ethylmorfolin, n-ethyl morpholine, n-ethylmorfolin czech, unii-ecm0g991fq, n-ethyl-morpholine, ccris 4818, ecm0g991fq | |
HVCNXQOWACZAFN-UHFFFAOYSA-N | |
4-ethylmorpholine | |
7525 | |
99% | |
N-Ethylmorpholine, 99% |
98.5 | |
-63.0°C | |
0.9000g/mL | |
27°C | |
98.5% min. (GC) | |
1.4400 to 1.4420 | |
27, 6 | |
0.9 | |
Solubility in water: miscible with water. Other solubilities: soluble in alcohol,ether,acetone and benzene | |
CCN1CCOCC1 | |
115.17 | |
115.17 | |
Powder |
Safety and Handling
GHS H Statement
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Harmful if inhaled.
Flammable liquid and vapor.
Harmful if swallowed.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 202-885-0
missing translation for 'rtecsNumber' : QE4025000
missing translation for 'tsca' : TSCA
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.