missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Ethylphthalimide, 98%, Thermo Scientific Chemicals
40.50 € - 465.00 €
Chemical Identifiers
CAS | 5022-29-7 |
---|---|
Molecular Formula | C10H9NO2 |
Molecular Weight (g/mol) | 175.187 |
MDL Number | MFCD00014583 |
InChI Key | JZDSOQSUCWVBMV-UHFFFAOYSA-N |
Synonym | n-ethylphthalimide, 2-ethylisoindoline-1,3-dione, phthalimide, n-ethyl, 1h-isoindole-1,3 2h-dione, 2-ethyl, 2-ethyl-1h-isoindole-1,3 2h-dione, 1h-isoindole-1,3 2h-dione, n-ethyl, 2-ethylbenzo c azoline-1,3-dione, acmc-1az90, 1h-isoindole-1, n-ethyl |
PubChem CID | 21120 |
IUPAC Name | 2-ethylisoindole-1,3-dione |
SMILES | CCN1C(=O)C2=CC=CC=C2C1=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11428636
|
Thermo Scientific Alfa Aesar
A16014.06 |
5 g |
40.50 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11438636
|
Thermo Scientific Alfa Aesar
A16014.14 |
25 g | |||||||
11448636
|
Thermo Scientific Alfa Aesar
A16014.22 |
100 g | |||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
5022-29-7 | |
175.187 | |
JZDSOQSUCWVBMV-UHFFFAOYSA-N | |
21120 | |
CCN1C(=O)C2=CC=CC=C2C1=O |
C10H9NO2 | |
MFCD00014583 | |
n-ethylphthalimide, 2-ethylisoindoline-1,3-dione, phthalimide, n-ethyl, 1h-isoindole-1,3 2h-dione, 2-ethyl, 2-ethyl-1h-isoindole-1,3 2h-dione, 1h-isoindole-1,3 2h-dione, n-ethyl, 2-ethylbenzo c azoline-1,3-dione, acmc-1az90, 1h-isoindole-1, n-ethyl | |
2-ethylisoindole-1,3-dione |
Specifications
5022-29-7 | |
C10H9NO2 | |
132094 | |
JZDSOQSUCWVBMV-UHFFFAOYSA-N | |
2-ethylisoindole-1,3-dione | |
21120 | |
98% |
74°C to 78°C | |
MFCD00014583 | |
n-ethylphthalimide, 2-ethylisoindoline-1,3-dione, phthalimide, n-ethyl, 1h-isoindole-1,3 2h-dione, 2-ethyl, 2-ethyl-1h-isoindole-1,3 2h-dione, 1h-isoindole-1,3 2h-dione, n-ethyl, 2-ethylbenzo c azoline-1,3-dione, acmc-1az90, 1h-isoindole-1, n-ethyl | |
CCN1C(=O)C2=CC=CC=C2C1=O | |
175.187 | |
175.19 | |
N-Ethylphthalimide |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 225-707-3
missing translation for 'rtecsNumber' : TI5075000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only