missing translation for 'onlineSavingsMsg'
Learn More

N-Ethylpropylamine, 97+%, Thermo Scientific Chemicals

80.10 € - 289.00 €

Chemical Identifiers

CAS 20193-20-8
Molecular Formula C5H13N
Molecular Weight (g/mol) 87.166
MDL Number MFCD00015198
InChI Key XCVNDBIXFPGMIW-UHFFFAOYSA-N
Synonym n-ethylpropylamine, 1-propanamine, n-ethyl, ethyl propyl amine, propylamine, n-ethyl, ethylpropylamine, n-ethyl-n-propylamine, ethyl-propyl-amine, n-propylethylamine, n-ethylpropanamine, ethyl-n-propylamine
PubChem CID 88398
IUPAC Name N-ethylpropan-1-amine
SMILES CCCNCC
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11377088
View Documents Promotion Details
Thermo Scientific Alfa Aesar
014846.06
5 g
80.10 €
5g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11387088
View Documents Promotion Details
Thermo Scientific Alfa Aesar
014846.14
25 g
289.00 €
25g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

20193-20-8
87.166
XCVNDBIXFPGMIW-UHFFFAOYSA-N
88398
CCCNCC
C5H13N
MFCD00015198
n-ethylpropylamine, 1-propanamine, n-ethyl, ethyl propyl amine, propylamine, n-ethyl, ethylpropylamine, n-ethyl-n-propylamine, ethyl-propyl-amine, n-propylethylamine, n-ethylpropanamine, ethyl-n-propylamine
N-ethylpropan-1-amine

Specifications

20193-20-8
80°C to 85°C
C5H13N
UN2733
XCVNDBIXFPGMIW-UHFFFAOYSA-N
N-ethylpropan-1-amine
88398
≥97%
N-Ethylpropylamine
0.748 g/mL
4.4°C (40°F)
MFCD00015198
n-ethylpropylamine, 1-propanamine, n-ethyl, ethyl propyl amine, propylamine, n-ethyl, ethylpropylamine, n-ethyl-n-propylamine, ethyl-propyl-amine, n-propylethylamine, n-ethylpropanamine, ethyl-n-propylamine
CCCNCC
87.166
87.16
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c

H225-H302+H332-H311-H314-H335

missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.

missing translation for 'einecsNumber' : 243-573-4

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only