Learn More
N-Fmoc-3-(4-pyridyl)-L-alanine, 95%, Thermo Scientific Chemicals
85.00 € - 1076.00 €
Chemical Identifiers
CAS | 169555-95-7 |
---|---|
Molecular Formula | C23H20N2O4 |
Molecular Weight (g/mol) | 388.423 |
MDL Number | MFCD00672566 |
InChI Key | SCSSXJVRZMQUKA-NRFANRHFSA-N |
Synonym | fmoc-l-4-pyridylalanine, fmoc-4-pal-oh, fmoc-3-4-pyridyl-l-alanine, fmoc-ala 4-pyridyl-oh, fmoc-beta-4-pyridyl-ala-oh, n-fmoc-3-4-pyridyl-l-alanine, fmoc-l-3-4-pyridyl-alanine, fmoc-4'-pyridyl-l-ala, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-pyridin-4-yl propanoic acid, fmoc-3-4-pyridyl-d-alanine |
PubChem CID | 978322 |
IUPAC Name | (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-pyridin-4-ylpropanoic acid |
SMILES | C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC4=CC=NC=C4)C(=O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15468717
|
Thermo Scientific Alfa Aesar
H52172.MD |
250 mg |
85.00 €
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15448717
|
Thermo Scientific Alfa Aesar
H52172.03 |
1 g |
262.00 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15458717
|
Thermo Scientific Alfa Aesar
H52172.06 |
5 g |
1076.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Fmoc-(4-pyridyl)-Ala-OH is potentially useful for solid phase peptide synthesis techniques.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsFmoc-∫-(4-pyridyl)-Ala-OH is potentially useful for solid phase peptide synthesis techniques.
Solubility
Soluble in 0.1g in 4mL DMF(clearly soluble).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents. Store at 4°C.
Chemical Identifiers
169555-95-7 | |
388.423 | |
SCSSXJVRZMQUKA-NRFANRHFSA-N | |
978322 | |
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC4=CC=NC=C4)C(=O)O |
C23H20N2O4 | |
MFCD00672566 | |
fmoc-l-4-pyridylalanine, fmoc-4-pal-oh, fmoc-3-4-pyridyl-l-alanine, fmoc-ala 4-pyridyl-oh, fmoc-beta-4-pyridyl-ala-oh, n-fmoc-3-4-pyridyl-l-alanine, fmoc-l-3-4-pyridyl-alanine, fmoc-4'-pyridyl-l-ala, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-pyridin-4-yl propanoic acid, fmoc-3-4-pyridyl-d-alanine | |
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-pyridin-4-ylpropanoic acid |
Specifications
169555-95-7 | |
MFCD00672566 | |
fmoc-l-4-pyridylalanine, fmoc-4-pal-oh, fmoc-3-4-pyridyl-l-alanine, fmoc-ala 4-pyridyl-oh, fmoc-beta-4-pyridyl-ala-oh, n-fmoc-3-4-pyridyl-l-alanine, fmoc-l-3-4-pyridyl-alanine, fmoc-4'-pyridyl-l-ala, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-pyridin-4-yl propanoic acid, fmoc-3-4-pyridyl-d-alanine | |
SCSSXJVRZMQUKA-NRFANRHFSA-N | |
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-pyridin-4-ylpropanoic acid | |
978322 | |
95% |
C23H20N2O4 | |
7316905 | |
Soluble in 0.1g in 4mL DMF(clearly soluble). | |
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC4=CC=NC=C4)C(=O)O | |
388.423 | |
388.42 | |
N-Fmoc-3-(4-pyridyl)-L-alanine |
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.