missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-L-glutamic acid 1-benzyl ester, 95%, Thermo Scientific Chemicals

Product Code. 15492779
5 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122350-52-1
Molecular Formula C27H25NO6
Molecular Weight (g/mol) 459.498
MDL Number MFCD00080272
InChI Key FMWLYDDRYGOYMY-DEOSSOPVSA-N
Synonym fmoc-glu-obzl, s-4-9h-fluoren-9-yl methoxy carbonyl amino-5-benzyloxy-5-oxopentanoic acid, 4s-5-benzyloxy-4-9h-fluoren-9-ylmethoxy carbonyl amino-5-oxopentanoic acid, fmoc-l-glutamic acid alpha-benzyl ester, nalpha-fmoc-l-glutamic acid alpha-benzyl ester
PubChem CID 10049854
IUPAC Name (4S)-4-(9H-fluoren-9-ylmethoxycarbonylamino)-5-oxo-5-phenylmethoxypentanoic acid
SMILES C1=CC=C(C=C1)COC(=O)C(CCC(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24

Specifications

Color White
Quantity 5 g
Formula Weight 459.5
Percent Purity 95%
Physical Form Powder
Chemical Name or Material N-Fmoc-L-glutamic acid 1-benzyl ester

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
N-Fmoc-L-glutamic acid 1-benzyl ester, 95%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.