missing translation for 'onlineSavingsMsg'
Learn More
N-Fmoc-O-benzyl-D-tyrosine, 95%, Thermo Scientific™ Product Code.: 15452789

N-Fmoc-O-benzyl-D-tyrosine, 95%, Thermo Scientific™

Product Code. 15452789
250 mg
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy

Product Code. 15452789

Brand: Thermo Scientific Alfa Aesar H66599.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 138775-48-1
Molecular Formula C31H27NO5
Molecular Weight (g/mol) 493.56
MDL Number MFCD00235837
InChI Key REHSJSKPWIOKIJ-KSTXGFNDNA-N
Synonym fmoc-d-tyr bzl-oh, fmoc-o-benzyl-d-tyr, r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-4-benzyloxy phenyl propanoic acid, 2r-3-4-benzyloxy phenyl-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid, fmoc-o-benzyl-l-tyr, n-alpha-9-fluorenylmethyloxycarbonyl-o-benzyl-d-tyrosine, o-benzyl-n-9h-fluoren-9-yl methoxy carbonyl-d-tyrosine, d-tyrosine,n-9h-fluoren-9-ylmethoxy carbonyl-o-phenylmethyl
PubChem CID 40428316
IUPAC Name (2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(4-phenylmethoxyphenyl)propanoic acid
SMILES OC(=O)[C@@H](CC1=CC=C(OCC2=CC=CC=C2)C=C1)NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2

Specifications

Color White
Quantity 250 mg
Formula Weight 439.56
Percent Purity 95%
Physical Form Powder
Chemical Name or Material N-Fmoc-O-benzyl-D-tyrosine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
N-Fmoc-O-benzyl-D-tyrosine, 95%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.