missing translation for 'onlineSavingsMsg'
Learn More
N-Fmoc-O-tert-butyl-N-methyl-L-threonine, 99%, Thermo Scientific Chemicals Product Code.: 15419718

N-Fmoc-O-tert-butyl-N-methyl-L-threonine, 99%, Thermo Scientific Chemicals

Product Code. 15419718
1 g
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 15419718

Brand: Thermo Scientific Alfa Aesar H63170.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 117106-20-4
Molecular Formula C24H29NO5
Molecular Weight (g/mol) 411.50
MDL Number MFCD02094431
InChI Key VIUVLZHFMIFLHU-VFNWGFHPSA-N
Synonym fmoc-n-me-thr tbu-oh, 2s,3r-2-9h-fluoren-9-yl methoxy carbonyl methyl amino-3-tert-butoxy butanoic acid, n-fmoc-n-methyl-o-tert-butyl-l-threonine, fmoc-n-methyl-o-t-butyl-l-threonine, pubchem19052, fmoc-n-me-thr but-oh, fmoc-nalpha-methyl-o-t-butyl-l-threonine, fmoc-n-alpha-methyl-o-t-butyl-l-threonine, n-alpha-9-fluorenylmethyloxycarbonyl-n-alpha-methyl-o-t-butyl-l-threonineShow MoreShow Less
PubChem CID 7010372
IUPAC Name (2S,3R)-2-[9H-fluoren-9-ylmethoxycarbonyl(methyl)amino]-3-[(2-methylpropan-2-yl)oxy]butanoic acid
SMILES C[C@@H](OC(C)(C)C)[C@H](N(C)C(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O

Specifications

Quantity 1 g
Formula Weight 411.49
Percent Purity 99%
Physical Form Solid
Chemical Name or Material N-Fmoc-O-tert-butyl-N-methyl-L-threonine

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberM28H042 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH63170.03
Lot Number81301454 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH63170.03

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
N-Fmoc-O-tert-butyl-N-methyl-L-threonine, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.