missing translation for 'onlineSavingsMsg'
Learn More

n-Heptadecane, 99%, Thermo Scientific Chemicals

Product Code. 11323907
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

n-Heptadecane is used as an internal standard to study the first direct comparison of kava lactone extraction efficiencies. It plays an important role as a suspended solvent used for the extraction and concentration of essential oil. It is used as a precursor in the production of iminoctadine triacetate

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 629-78-7
Molecular Formula C17H36
Molecular Weight (g/mol) 240.475
MDL Number MFCD00009002
InChI Key NDJKXXJCMXVBJW-UHFFFAOYSA-N
Synonym n-heptadecane, heptadekan, unii-h7c0j39xum, hexadecane, methyl, h7c0j39xum, heptadecane, analytical standard, normal-heptadecane, heptadecane, dsstox_cid_27061, dsstox_rid_82078
PubChem CID 12398
ChEBI CHEBI:16148
IUPAC Name heptadecane
SMILES CCCCCCCCCCCCCCCCC

Specifications

Melting Point 22°C
Density 0.778 g/mL
Boiling Point 302°C
Refractive Index 1.4369
Quantity 500 g
Solubility Information Immiscible with water.
Formula Weight 240.48
Percent Purity 99%
Physical Form Low Melting Solid
Chemical Name or Material n-Heptadecane
compliance-icons
Product Identifier
  • n-Heptadecane
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
Supplemental information
  • MIXTURE LIST-Contains : Heptadecane

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'