missing translation for 'onlineSavingsMsg'
Learn More
Learn More
n-Heptane, 99.5%, for analysis
Tous les produits Thermo Scientific Chemicals Produits
Click to view available options
Quantity:
1 L
2.5 L
25 L
Conditionnement:
1 L
2.5 L
25 L
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For analysis
Spécification
Spécification
| Melting Point | -91.0°C |
| CAS Min % | 99.4 |
| Acidity | 0.0001 meq/g max. |
| CAS Max % | 100.0 |
| Density | 0.6830g/mL |
| Boiling Point | 98.0°C |
| Quantity | 1 L |
| Flash Point | −4°C |
| Packaging | Glass Bottle |
| Physical Form | Liquid |
| Afficher plus |
Sécurité et traitement
Identifiant produit
- n-Heptane
- Danger
- Flammable liquid Category 2
- Aspiration toxicity Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
- H225-Highly flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H336-May cause drowsiness or dizziness.
- H410-Very toxic to aquatic life with long lasting effects.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P331-Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P273-Avoid release to the environment.
RUO – Research Use Only
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu