missing translation for 'onlineSavingsMsg'
Learn More

n-Heptane, 99+%, extra pure, Thermo Scientific Chemicals

Product Code. 10388402
Click to view available options
Quantity:
1 L
2.5 L
5 L
25 L
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142-82-5
Molecular Formula C7H16
Molecular Weight (g/mol) 100.21
MDL Number MFCD00009544
InChI Key IMNFDUFMRHMDMM-UHFFFAOYSA-N
Synonym n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish
PubChem CID 8900
ChEBI CHEBI:43098
IUPAC Name heptane
SMILES CCCCCCC

Specifications

Melting Point -91.0°C
Color Colorless to Yellow
Acidity 0.003 meq/g max.
Density 0.6830g/mL
Boiling Point 98.0°C
Quantity 5 L
Flash Point −4°C
Infrared Spectrum Authentic
Packaging Plastic Drum
Physical Form Liquid
Refractive Index 1.3850 to 1.3890
Assay Percent Range 99+%
Residue after Evaporation 20ppm max.
Specific Gravity 0.683
Linear Formula CH3(CH2)5CH3
Viscosity 0.4 mPa.s (20°C)
Water 0.05% max.
Merck Index 15, 4694
Solubility Information Solubility in water: practically insoluble. Other solubilities: soluble in alcohol, ether and chloroform
Formula Weight 100.2
Percent Purity ≥99%
Grade Extra Pure
Chemical Name or Material n-Heptane
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • n-Heptane
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Aspiration toxicity Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P273-Avoid release to the environment.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'