missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Heptane, Certified AR for Analysis, Fisher Chemical™
108.71 EUR valid until 2024-12-31
Use promo code "23271" to get your promotional price.
Use promo code "23271" to get your promotional price.
Click to view available options
Quantity:
500 mL
1 L
2.5 L
5 L
Packaging:
Amber Glass Bottle
Metal Can
Plastic Container
Description
C7H16, CAS Number-142-82-5, dipropylmethane, dipropyl methane, eptani, n-heptane, gettysolve-c, heptan polish, skellysolve c, heptyl hydride, heptan, heptanen, 2.5L, 98 deg.C, CHEBI:43098, Colorless, 100.2g/mol, IMNFDUFMRHMDMM-UHFFFAOYSA-N, -91 deg.C, 9544, 100.205, Amber glass bottle, Liquid, 8900
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | -91°C |
Color | Colorless |
Boiling Point | 98°C |
Quantity | 2.5 L |
Packaging | Amber Glass Bottle |
Physical Form | Liquid |
Vapor Pressure | 48mbar at 20°C |
Viscosity | 0.4 mPaS at 20°C |
Formula Weight | 100.2g/mol |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS02_Extremely_Flammable.png)
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS07_Warning.png)
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS08_Dangerous_Health_Hazard.png)
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS09_Aquatic_Toxicity.png)
Product Identifier
- n-Heptane
- Danger
- Flammable liquid Category 2
- Aspiration toxicity Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
- H225-Highly flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H336-May cause drowsiness or dizziness.
- H410-Very toxic to aquatic life with long lasting effects.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P331-Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P273-Avoid release to the environment.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'