Learn More
n-Hexadecane, 99%, Thermo Scientific Chemicals
48.24 € - 523.61 €
Chemical Identifiers
CAS | 544-76-3 |
---|---|
Molecular Formula | C16H34 |
Molecular Weight (g/mol) | 226.448 |
MDL Number | MFCD00008998 |
InChI Key | DCAYPVUWAIABOU-UHFFFAOYSA-N |
Synonym | n-hexadecane, cetane, n-cetane, hexadecan, hexadekan, cetan, zetan, unii-f8z00shp6q, ccris 5833, pentadecane, methyl |
PubChem CID | 11006 |
ChEBI | CHEBI:45296 |
IUPAC Name | hexadecane |
SMILES | CCCCCCCCCCCCCCCC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11414093
|
Thermo Scientific Alfa Aesar
A10322.AE |
100 mL |
48.24 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11424093
|
Thermo Scientific Alfa Aesar
A10322.AP |
500 mL |
160.80 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11434093
|
Thermo Scientific Alfa Aesar
A10322.0F |
2500 mL |
523.61 €
2500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Hexadecane is use in the recovery of valuable amines by means of distillation from aqueous alkanolamine solutions, which are extensively used in the gas-processing industries.1
1Abdi, M. A.; Meisen, A. Mutual Solubility of Hexadecane with Diethanolamine and with Bis(hydroxyethyl)piperazine. J. Chem. Eng. Data 1998, 43 (2), 138-142.
n-Hexadecane serves as a reference for other fuel mixtures and is assigned cetane number of 100; used as shorthand for cetane number, an indicator of the combustion speed of diesel fuel. Hexadecane is used in the preparation of emulsions; also used as a substrate for the production of biosurfactants. n-Hexadecane has been found useful in the identification of the wheat seed protein CM3 as a highly active emulsifier. Superamphiphobicity on polymers can be studied with hexadecane immersion tests. Shape-stabilized phase change material (SSPCM), prepared using hexadecane as a PCM, into xGnP as a supporting material, has been demonstrated to have the potential to be used in developing building materials with improved thermal energy storage charecteristics.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsHexadecane is use in the recovery of valuable amines by means of distillation from aqueous alkanolamine solutions, which are extensively used in the gas-processing industries.1
1Abdi, M. A.; Meisen, A. Mutual Solubility of Hexadecane with Diethanolamine and with Bis(hydroxyethyl)piperazine. J. Chem. Eng. Data 1998, 43 (2), 138-142.n-Hexadecane serves as a reference for other fuel mixtures and is assigned cetane number of 100; used as shorthand for cetane number, an indicator of the combustion speed of diesel fuel. Hexadecane is used in the preparation of emulsions; also used as a substrate for the production of biosurfactants. n-Hexadecane has been found useful in the identification of the wheat seed protein CM3 as a highly active emulsifier. Superamphiphobicity on polymers can be studied with hexadecane immersion tests. Shape-stabilized phase change material (SSPCM), prepared using hexadecane as a PCM, into xGnP as a supporting material, has been demonstrated to have the potential to be used in developing building materials with improved thermal energy storage charecteristics.
Notes
Stable. Cetane can ignite very easily under compression. Incompatible with strong oxidizing agents.
Chemical Identifiers
544-76-3 | |
226.448 | |
DCAYPVUWAIABOU-UHFFFAOYSA-N | |
11006 | |
hexadecane |
C16H34 | |
MFCD00008998 | |
n-hexadecane, cetane, n-cetane, hexadecan, hexadekan, cetan, zetan, unii-f8z00shp6q, ccris 5833, pentadecane, methylShow More | |
CHEBI:45296 | |
CCCCCCCCCCCCCCCC |
Specifications
544-76-3 | |
0.773 | |
135°C (275°F) | |
1.4345 | |
1736592 | |
This product is soluble in acetone,alcohol,ether. It is insoluble in water.Soluble in alcohol,acetone,chloroform,and ether. insoluble in water | |
CCCCCCCCCCCCCCCC | |
226.448 | |
CHEBI:45296 | |
n-Hexadecane |
18°C | |
287°C | |
C16H34 | |
MFCD00008998 | |
n-hexadecane, cetane, n-cetane, hexadecan, hexadekan, cetan, zetan, unii-f8z00shp6q, ccris 5833, pentadecane, methyl | |
DCAYPVUWAIABOU-UHFFFAOYSA-N | |
hexadecane | |
11006 | |
226.45 |
Safety and Handling
P301+P310-P331-P501c
H304
missing translation for 'einecsNumber' : 208-878-9
missing translation for 'rtecsNumber' : ML9200000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- n-Hexadecane
Signal Word
- Danger
Hazard Category
- Aspiration toxicity Category 1
Hazard Statement
- H304-May be fatal if swallowed and enters airways.
- EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P331-Do NOT induce vomiting.
Supplemental information
- MIXTURE LIST-Contains : Hexadecane
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.