Learn More
n-Hexane, 95+%, ACS reagent, Thermo Scientific Chemicals
69.55 € - 145.73 €
Chemical Identifiers
CAS | 110-54-3 |
---|---|
Molecular Formula | C6H14 |
Molecular Weight (g/mol) | 86.18 |
MDL Number | MFCD02179311 |
InChI Key | VLKZOEOYAKHREP-UHFFFAOYSA-N |
IUPAC Name | hexane |
SMILES | CCCCCC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
12387163
|
Thermo Scientific Acros
445780010 |
1 L |
69.55 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12307173
|
Thermo Scientific Acros
445780025 |
2.5 L |
145.73 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
110-54-3 | |
86.18 | |
VLKZOEOYAKHREP-UHFFFAOYSA-N | |
CCCCCC |
C6H14 | |
MFCD02179311 | |
hexane |
Specifications
-95.0°C | |
0.6590 g/mL | |
-22°C | |
95+% | |
C6H14 | |
MFCD02179311 | |
15, 4732 | |
VLKZOEOYAKHREP-UHFFFAOYSA-N | |
hexane | |
86.18 | |
n-Hexane |
110-54-3 | |
69.0°C | |
1.3748 to 1.3810 | |
0.659 | |
CH3(CH2)4CH3 | |
0.31 mPa.s (20°C) | |
Solubility aq. soln.: insoluble. Other solubilities: soluble in alcohol, acetone, ether and chloroform | |
CCCCCC | |
86.18 | |
ACS Reagent |
Safety and Handling
GHS H Statement May be fatal if swallowed and enters airways.
May cause drowsiness or dizziness.
Causes skin irritation.
May cause damage to organs through prolonged or repeated exposure.
Toxic to aquatic life with long las
GHS P Statement Keep away from heat, hot surfaces, sparks, open flames and other ignition sources.
No smoking.
Use personal protective equipment as required.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 203-777-6
- n-Hexane
Signal Word
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 2
- Aspiration toxicity Category 1
- Flammable liquid Category 2
- Reproductive toxicity Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity after repeated exposure Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H336-May cause drowsiness or dizziness.
- H361f-Suspected of damaging fertility.
- H373-May cause damage to organs.
- H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P331-Do NOT induce vomiting.
Supplemental information
- MIXTURE LIST-Contains : Hexane
RUO – Research Use Only