Learn More
n-Hexane, 96+%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
30.45 € - 296.48 €
Chemical Identifiers
CAS | 110-54-3 |
---|---|
Molecular Formula | C6H14 |
Molecular Weight (g/mol) | 86.18 |
MDL Number | MFCD02179311 |
InChI Key | VLKZOEOYAKHREP-UHFFFAOYSA-N |
Synonym | n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes |
PubChem CID | 8058 |
ChEBI | CHEBI:29021 |
IUPAC Name | hexane |
SMILES | CCCCCC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10061871
|
Thermo Scientific Acros
326921000 |
100 mL |
30.45 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10292412
|
Thermo Scientific Acros
326920010 |
1 L |
144.72 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10359233
|
Thermo Scientific Acros
326920025 |
2.5 L |
296.48 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
110-54-3 | |
86.18 | |
VLKZOEOYAKHREP-UHFFFAOYSA-N | |
8058 | |
hexane |
C6H14 | |
MFCD02179311 | |
n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes | |
CHEBI:29021 | |
CCCCCC |
Specifications
-95.0°C | |
96.0 | |
100.0 | |
69.0°C | |
AcroSeal™ Glass Bottle | |
1.3748 to 1.3810 | |
0.0005 % max. | |
C6H14 | |
MFCD02179311 | |
01, 142 | |
15, 4732 | |
Solubility in water: insoluble. Other solubilities: soluble in alcohol, acetone, ether and chloroform | |
CCCCCC | |
86.18 | |
CHEBI:29021 | |
≥96% | |
n-Hexane |
110-54-3 | |
Colorless | |
0.6590g/mL | |
−22°C | |
Liquid | |
96+% | |
0.659 | |
CH3(CH2)4CH3 | |
0.31 mPa.s (20°C) | |
0.002% Max. | |
n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes | |
VLKZOEOYAKHREP-UHFFFAOYSA-N | |
hexane | |
8058 | |
86.18 | |
Extra Dry |
Safety and Handling
GHS P Statement May be fatal if swallowed and enters airways. May cause drowsiness or dizziness. Causes skin irritation. May cause damage to organs through prolonged or repeated exposure. Toxic to aquatic life with long lasting effects. Suspected of damaging fertility. Highly flammable liquid and vapor.
GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Use personal protective equipment as required. Do not breathe dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician. Do NOT induce vomiting. Avoid release to the environment. IF ON SKIN: Wash with plenty of soap and water.
Danger
- n-Hexane
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Aspiration toxicity Category 1
- Skin corrosion/irritation Category 2
- Reproductive toxicity Category 2
- Specific target organ toxicity Category 3
- Specific target organ toxicity after repeated exposure Category 2
- LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H336-May cause drowsiness or dizziness.
- H361f-Suspected of damaging fertility.
- H373-May cause damage to organs.
- H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P331-Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P273-Avoid release to the environment.
RUO – Research Use Only