missing translation for 'onlineSavingsMsg'
Learn More

n-Hexane, 97%, for electronic use (MOS), residue free, Thermo Scientific Chemicals

Product Code. 10469062
Click to view available options
Quantity:
1 L
Packaging:
Glass Bottle
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Melting Point -95.0°C
CAS 110-54-3
CAS Min % 96.0
Acidity 0.002% max.
CAS Max % 100.0
Density 0.6590g/mL
Boiling Point 69.0°C
Quantity 1 L
Flash Point −22°C
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.3748 to 1.3810
Assay Percent Range 97%
Residue after Evaporation 0.0005% max.
Specific Gravity 0.659
Molecular Formula C6H14
Linear Formula CH3(CH2)4CH3
MDL Number MFCD02179311
UN Number 1208
Viscosity 0.31 mPa.s (20°C)
Beilstein 01, 142
Water 0.01% max. (Coulometric)
Merck Index 15, 4732
Synonym n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol, acetone, ether and chloroform
InChI Key VLKZOEOYAKHREP-UHFFFAOYSA-N
SMILES CCCCCC
IUPAC Name hexane
Molecular Weight (g/mol) 86.18
PubChem CID 8058
ChEBI CHEBI:29021
Formula Weight 86.18
Percent Purity 97%
Grade Electronic
Chemical Name or Material n-Hexane, Residue Free
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • n-Hexane
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Aspiration toxicity Category 1
  • Skin corrosion/irritation Category 2
  • Reproductive toxicity Category 2
  • Specific target organ toxicity Category 3
  • Specific target organ toxicity after repeated exposure Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H361f-Suspected of damaging fertility.
  • H373-May cause damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P273-Avoid release to the environment.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'