missing translation for 'onlineSavingsMsg'
Learn More

n-Hexane, anhydrous, over molecular sieves, packaged under argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals

Product Code. 15465065
Click to view available options
Quantity:
250 mL
1 L
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

n-Hexane is used as a solvent, paint thinner, and chemical reaction medium. It is used to extract edible oils from seeds and vegetables, as a special-use solvent, and as a cleaning agent. Hexanes are commonly used in chromatography as a non-polar solvent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-54-3
Molecular Formula C6H14
Molecular Weight (g/mol) 86.18
MDL Number MFCD02179311
InChI Key VLKZOEOYAKHREP-UHFFFAOYSA-N
Synonym n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
PubChem CID 8058
ChEBI CHEBI:29021
IUPAC Name hexane
SMILES CCCCCC

Specifications

Melting Point -95°C
Color Colorless
Density 0.659 g/mL
Boiling Point 69°C
Quantity 250 mL
Flash Point -23°C (-9°F)
Packaging ChemSeal™ Bottle
Physical Form Liquid
Refractive Index 1.375
Linear Formula CH3(CH2)4CH3
UN Number UN1208
Beilstein 1730733
Merck Index 14,4694
Solubility Information Insoluble in water
Formula Weight 86.18
Chemical Name or Material n-Hexane, Anhydrous, over molecular sieves
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'