missing translation for 'onlineSavingsMsg'
Learn More

n-Hexane, HPLC Grade, 95% min, Thermo Scientific Chemicals

79.50 € - 220.10 €

Chemical Identifiers

CAS 110-54-3
Molecular Formula C6H14
Molecular Weight (g/mol) 86.18
MDL Number MFCD02179311
InChI Key VLKZOEOYAKHREP-UHFFFAOYSA-N
Synonym n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
PubChem CID 8058
ChEBI CHEBI:29021
IUPAC Name hexane
SMILES CCCCCC
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11363588
View Documents
Thermo Scientific Alfa Aesar
039199.K2
1 L
79.50 €
1 L
Please to purchase this item. Need a web account? Register with us today!
11373588
View Documents
Thermo Scientific Alfa Aesar
039199.M1
4 x 1 L
220.10 €
4 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

as a solvent to extract edible oils from seed and vegetable crops. Commercial grades of hexane are used as solvents for glues, varnishes, and inks, cleaning agent (degreaser) in the printing industry, liquid in low temperature thermometers. other uses are as Fuels and fuel additives, Intermediates and Laboratory chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

110-54-3
86.18
VLKZOEOYAKHREP-UHFFFAOYSA-N
8058
hexane
C6H14
MFCD02179311
n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
CHEBI:29021
CCCCCC

Specifications

°C to 95°C
0.659
-23°C (-9°F)
1.375
C6H14
1730733
Very soluble in ethanol,ethyl ether and chloroform.
CCCCCC
86.18
CHEBI:29021
HPLC
110-54-3
69°C
Liquid, Distilled in Glass
95% min
MFCD02179311
n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
VLKZOEOYAKHREP-UHFFFAOYSA-N
hexane
8058
86.18
n-Hexane
Videos
Safety and Handling

Safety and Handling

P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280i-P281-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P331-P332+P313-P363-P370+P378q-P501c

H225-H304-H315-H319-H335-H336-H361f-H372

missing translation for 'dotInformation' : Hazard Class: 3; Packaging Group: II

missing translation for 'einecsNumber' : 203-777-6

missing translation for 'rtecsNumber' : MN9275000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Product Identifier
  • n-Hexane, HPLC Grade

Signal Word
  • Danger

Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Aspiration toxicity Category 1
  • Flammable liquid Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H361f-Suspected of damaging fertility.
  • H373-May cause damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P331-Do NOT induce vomiting.

Supplemental information
  • MIXTURE LIST-Contains : hexane

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.