missing translation for 'onlineSavingsMsg'
Learn More

n-Hexane, Spectrophotometric Grade, 95+%, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals

30.35 € - 62.91 €

Chemical Identifiers

CAS 110-54-3
Molecular Formula C6H14
Molecular Weight (g/mol) 86.18
MDL Number MFCD02179311
InChI Key VLKZOEOYAKHREP-UHFFFAOYSA-N
Synonym n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
PubChem CID 8058
ChEBI CHEBI:29021
IUPAC Name hexane
SMILES CCCCCC
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11318888
View Documents
Thermo Scientific Alfa Aesar
041727.AH
200 mL
30.35 €
200mL
Please to purchase this item. Need a web account? Register with us today!
11308888
View Documents
Thermo Scientific Alfa Aesar
041727.9Z
800 mL
62.91 €
800mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

n-Hexane is used in the formulation of glues for shoes, leather products, and roofing, as a cleaning agent, de-greasing agent. It is used to extract edible oils from seeds and vegetables, a non-polar solvent in chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
Specifications

Chemical Identifiers

110-54-3
86.18
VLKZOEOYAKHREP-UHFFFAOYSA-N
8058
hexane
C6H14
MFCD02179311
n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
CHEBI:29021
CCCCCC

Specifications

-95°C
0.659 g/mL
-23°C (-9°F)
ChemSeal™ Bottle
1.375
MFCD02179311
1730733
n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
VLKZOEOYAKHREP-UHFFFAOYSA-N
hexane
8058
86.18
Spectrophotometric
110-54-3
69°C
Weak, characteristic
Liquid
C6H14
UN1208
14,4694
Soluble in ethyl ether, chloroform. Miscible with ethanol.
CCCCCC
86.18
CHEBI:29021
≥95%
n-Hexane
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H304-H361-H373-H315-H336
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Causes skin irritation.
May cause drowsiness or dizziness.

P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280i-P281-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P331-P332+P313-P363-P370+P378q-P501c

H225-H304-H315-H319-H335-H336-H361f-H372

missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HEXANES

missing translation for 'einecsNumber' : 203-777-6

missing translation for 'rtecsNumber' : MN9275000

missing translation for 'tsca' : Yes

missing translation for 'storageNote' :

  • Packaged under argon

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only