Learn More
n-Hexane, Spectrophotometric Grade, 95+%, Thermo Scientific Chemicals
90.70 € - 281.00 €
Chemical Identifiers
CAS | 110-54-3 |
---|---|
Molecular Formula | C6H14 |
Molecular Weight (g/mol) | 86.18 |
MDL Number | MFCD02179311 |
InChI Key | VLKZOEOYAKHREP-UHFFFAOYSA-N |
Synonym | n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes |
PubChem CID | 8058 |
ChEBI | CHEBI:29021 |
IUPAC Name | hexane |
SMILES | CCCCCC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11338397
|
Thermo Scientific Alfa Aesar
032454.K2 |
1 L |
90.70 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11348397
|
Thermo Scientific Alfa Aesar
032454.M1 |
4 x 1 L |
281.00 €
4 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
n-Hexane used in spectrophotometry and environmental testing.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
110-54-3 | |
86.18 | |
VLKZOEOYAKHREP-UHFFFAOYSA-N | |
8058 | |
hexane |
C6H14 | |
MFCD02179311 | |
n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes | |
CHEBI:29021 | |
CCCCCC |
Specifications
-95°C | |
0.659 | |
−23°C (−9°F) | |
Liquid | |
C6H14 | |
UN1208 | |
14,4694 | |
Very soluble in ethanol,ethyl ether and chloroform. | |
CCCCCC | |
86.18 | |
CHEBI:29021 | |
≥95% | |
n-Hexane |
110-54-3 | |
69°C | |
Weak, Characteristic | |
1.375 | |
MFCD02179311 | |
1730733 | |
n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes | |
VLKZOEOYAKHREP-UHFFFAOYSA-N | |
hexane | |
8058 | |
86.18 | |
Spectrophotometric |
Safety and Handling
GHS H Statement
H225-H304-H361-H373-H315-H336;
Highly flammable liquid and vapour.
May be fatal if swallowed and enters airways.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Causes skin irritation.
May cause drowsiness or dizziness.
P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280i-P281-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P331-P332+P313-P363-P370+P378q-P501c
H225-H304-H315-H319-H335-H336-H361f-H372
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HEXANES
missing translation for 'einecsNumber' : 203-777-6
missing translation for 'rtecsNumber' : MN9275000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only