missing translation for 'onlineSavingsMsg'
Learn More

n-Hexyl methacrylate, 97%, stab. with 100ppm 4-methoxyphenol, Thermo Scientific Chemicals

Product Code. 11312507
100 g
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142-09-6
Molecular Formula C10H18O2
Molecular Weight (g/mol) 170.25
MDL Number MFCD00015283
InChI Key LNCPIMCVTKXXOY-UHFFFAOYSA-N
Synonym hexyl methacrylate, n-hexyl methacrylate, hexyl 2-methyl-2-propenoate, methacrylic acid, hexyl ester, 2-propenoic acid, 2-methyl-, hexyl ester, unii-538b2q3hsv, ccris 4824, hexyl 2-methylacrylate, methacrylic acid hexyl ester, dsstox_cid_5406
PubChem CID 8872
IUPAC Name hexyl 2-methylprop-2-enoate
SMILES CCCCCCOC(=O)C(C)=C

Specifications

Melting Point -57°C
Density 0.885
Boiling Point 204°C
Flash Point 80°C (176°F)
Odor Characteristic
Refractive Index 1.431
Quantity 100 g
Beilstein 1754703
Formula Weight 170.25
Percent Purity 97%
Chemical Name or Material n-Hexyl methacrylate, stabilized with 100ppm 4-methoxyphenol
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
n-Hexyl methacrylate, 97%, stab. with 100ppm 4-methoxyphenol, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.