missing translation for 'onlineSavingsMsg'
Learn More
n-Hexylbenzene, 98%, Thermo Scientific Chemicals Product Code.: 11415203

n-Hexylbenzene, 98%, Thermo Scientific Chemicals

Product Code. 11415203
25 g
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 11415203

Brand: Thermo Scientific Alfa Aesar A12821.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1077-16-3
Molecular Formula C12H18
Molecular Weight (g/mol) 162.28
MDL Number MFCD00009526
InChI Key LTEQMZWBSYACLV-UHFFFAOYSA-N
Synonym 1-phenylhexane, n-hexylbenzene, benzene, hexyl, hexane, 1-phenyl, unii-8e3d1a2bzp, 8e3d1a2bzp, phenylhexane, hexyl-benzen, 1-hexylbenzene, 4-hexylbenzene
PubChem CID 14109
IUPAC Name hexylbenzene
SMILES CCCCCCC1=CC=CC=C1

Specifications

Melting Point -61°C
Density 0.861
Boiling Point 226°C to 227°C
Flash Point 83°C (181°F)
Refractive Index 1.486
Quantity 25 g
UN Number UN3082
Beilstein 1904786
Formula Weight 162.28
Percent Purity 98%
Chemical Name or Material n-Hexylbenzene
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10252181 Certificate TypeCertificate of Analysis Date23/08/2024 Catalog NumberA12821.22,A12821.14,A12821.36
Lot Number10200382 Certificate TypeCertificate of Analysis Date29/12/2023 Catalog NumberA12821.22,A12821.14,A12821.36
Lot Number10206925 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA12821.22,A12821.14,A12821.36
Lot Number10217086 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA12821.22,A12821.14,A12821.36
Lot Number10185885 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA12821.22,A12821.14,A12821.36

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
n-Hexylbenzene, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.