missing translation for 'onlineSavingsMsg'
Learn More
n-Hexyllithium, 2.5M (33 wt.%) solution in hexane, AcroSeal™, Thermo Scientific Chemicals Product Code.: 10115320

n-Hexyllithium, 2.5M (33 wt.%) solution in hexane, AcroSeal™, Thermo Scientific Chemicals

Product Code. 10115320
800 mL, AcroSeal™ Glass Bottle
Click to view available options
Quantity:
800 mL
Packaging:
AcroSeal™ Glass Bottle
This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 
Sea freight only: For EU stock allow up to 15 days delivery.
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

Product Code. 10115320

Brand: Thermo Scientific Acros 301658000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 
Sea freight only: For EU stock allow up to 15 days delivery.
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21369-64-2, 110-54-3
Molecular Formula C6H13Li
Molecular Weight (g/mol) 92.11
MDL Number MFCD00191446
InChI Key CETVQRFGPOGIQJ-UHFFFAOYSA-N
Synonym n-hexyllithium, lithium, hexyl, hexyllithium, hexyl lithium, hexyl-lithium, lithium hexane, n-hexyllithiurn, n-hexyllitium, n-hexyl lithium, hexli
PubChem CID 2733163
IUPAC Name lithium;hexane
SMILES [Li+].CCCCC[CH2-]

Specifications

CAS Min % 32.8, 66.8
CAS Max % 33.2, 67.2
Color Orange to Yellow
Density 0.6900g/mL
Flash Point −26°C
Packaging AcroSeal™ Glass Bottle
Linear Formula LiCH2CH2CH2CH2CH2CH3
Quantity 800 mL
Specific Gravity 0.69
Solubility Information Solubility in water: vigorous reaction.
Formula Weight 92.11
Percent Purity 32.8 to 33.2% (as HexLi)
Physical Form Solution
Chemical Name or Material n-Hexyllithium, 33 wt. % in n-hexane
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0467896 Certificate TypeCertificate of Analysis Date12/02/2025 Catalog Number301651000,10115320,301658000
Lot NumberA0465772 Certificate TypeCertificate of Analysis Date05/09/2024 Catalog Number301651000,301658000
Lot NumberA0451421 Certificate TypeCertificate of Analysis Date04/01/2024 Catalog Number301651000,301658000
Lot NumberA0433390 Certificate TypeCertificate of Analysis Date06/10/2023 Catalog Number301651000,301658000
Lot NumberA0455963 Certificate TypeCertificate of Analysis Date28/08/2023 Catalog Number301651000,301658000
compliance-icons compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • n-Hexyllithium, 33 wt.% solution in n-hexane
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Substance which\, in contact with water\, emits flammable gases Category 1
  • Pyrophoric liquid Category 1
  • Aspiration toxicity Category 1
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
  • Reproductive toxicity Category 2
  • Specific target organ toxicity Category 3
  • Specific target organ toxicity after repeated exposure Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H250-Catches fire spontaneously if exposed to air.
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • H304-May be fatal if swallowed and enters airways.
  • H314-Causes severe skin burns and eye damage.
  • H336-May cause drowsiness or dizziness.
  • H361f-Suspected of damaging fertility.
  • H373-May cause damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Hydrocarbons, C6, n-alkanes, isoalkanes,cyclics, 5-50% n-hexane, n-Hexyllithium

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
n-Hexyllithium, 2.5M (33 wt.%) solution in hexane, AcroSeal™, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.