missing translation for 'onlineSavingsMsg'
Learn More

N-Hydroxy-2-methylpropanimidamide, 97%, Thermo Scientific™

Product Code. 10085062
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 35613-84-4
Molecular Formula C4H10N2O
Molecular Weight (g/mol) 102.14
MDL Number MFCD03426251,MFCD08705910,MFCD00464225
InChI Key JHRDEHLFNLLCQS-UHFFFAOYSA-N
Synonym z-n'-hydroxyisobutyrimidamide, 1z-n'-hydroxy-2-methylpropanimidamide, n-hydroxyisobutyrimidamide, n-hydroxy-2-methylpropanimidamide, z-n'-hydroxy-2-methylpropimidamide, n-hydroxyisobutyramidine, z-n'-hydroxy-2-methylpropanimidamide, isobutyramidoxime 250mg, n/'-hydroxy-2-methylpropanimidamide, propanimidamide, n-hydroxy-2-methyl
PubChem CID 9582835
IUPAC Name N'-hydroxy-2-methylpropanimidamide
SMILES CC(C)C(\N)=N/O

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 102.14
Percent Purity 97%
Chemical Name or Material N';-Hydroxy-2-methylpropanimidamide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.