missing translation for 'onlineSavingsMsg'
Learn More

N'-Hydroxy-2-phenylethanimidamide ≥97%, Thermo Scientific™

Product Code. 10471273
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
missing translation for 'unitSize'
10g
1g
This item is not returnable. View return policy

Product Code. 10471273

missing translation for 'mfr': Thermo Scientific Maybridge S01215EA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 19227-11-3
Molecular Formula C8H10N2O
Molecular Weight (g/mol) 150.181
MDL Number MFCD00019953
InChI Key FVYBAJYRRIYNBN-UHFFFAOYSA-N
Synonym 2-phenylacetamidoxime, n-hydroxy-2-phenylacetimidamide, 1z-n'-hydroxy-2-phenylethanimidamide, n-hydroxy-2-phenyl-acetamidine, z-n'-hydroxy-2-phenylacetimidamide, 1-hydroxyimino-2-phenylethylamine, n-hydroxy-2-phenylacetamidine, benzeneethanimidamide,n-hydroxy, benzeneethanimidamide, n-hydroxy, z-n'-hydroxy-2-phenylethanimidamide
PubChem CID 9566465
IUPAC Name N'-hydroxy-2-phenylethanimidamide
SMILES C1=CC=C(C=C1)CC(=NO)N

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 150.18
Percent Purity 97%
Chemical Name or Material N'-hydroxy-2-phenylethanimidamide
compliance-icons
Product Identifier
  • N'-Hydroxy-2-phenylethanimidamide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.