missing translation for 'onlineSavingsMsg'
Learn More

N-Hydroxysuccinimide, 98+%, Thermo Scientific Chemicals

38.59 € - 809.03 €

Chemical Identifiers

CAS 6066-82-6
Molecular Formula C4H5NO3
Molecular Weight (g/mol) 115.09
MDL Number MFCD00005516
InChI Key NQTADLQHYWFPDB-UHFFFAOYSA-N
Synonym n-hydroxysuccinimide, hosu, 1-hydroxysuccinimide, 2,5-pyrrolidinedione, 1-hydroxy, 1-hydroxy-2,5-pyrrolidinedione, succinimide, n-hydroxy, n-hydroxysuccinimde, n-hydroxysuccinimid, unii-mje3791m4t, ccris 2604
PubChem CID 80170
IUPAC Name 1-hydroxypyrrolidine-2,5-dione
SMILES C1CC(=O)N(C1=O)O
View More Specs

Products 4
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10255580
View Documents Promotion Details
Thermo Scientific Acros
157270250
25 g Plastic bottle
38.59 €
25g
Please to purchase this item. Need a web account? Register with us today!
10437940
View Documents Promotion Details
Thermo Scientific Acros
157271000
100 g Plastic bottle
110.55 €
100g
Please to purchase this item. Need a web account? Register with us today!
10581211
View Documents Promotion Details
Thermo Scientific Acros
157272500
250 g Plastic bottle
250.25 €
250g
Please to purchase this item. Need a web account? Register with us today!
10052973
View Documents Promotion Details
Thermo Scientific Acros
157270010
1 kg Plastic bottle
809.03 €
1kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent
Specifications

Chemical Identifiers

6066-82-6
115.09
NQTADLQHYWFPDB-UHFFFAOYSA-N
80170
C1CC(=O)N(C1=O)O
C4H5NO3
MFCD00005516
n-hydroxysuccinimide, hosu, 1-hydroxysuccinimide, 2,5-pyrrolidinedione, 1-hydroxy, 1-hydroxy-2,5-pyrrolidinedione, succinimide, n-hydroxy, n-hydroxysuccinimde, n-hydroxysuccinimid, unii-mje3791m4t, ccris 2604
1-hydroxypyrrolidine-2,5-dione

Specifications

6066-82-6
100.0
Brown to Brown-Yellow
Authentic
C4H5NO3
21, 380
15, 4884
which is virtually insoluble in acetone
C1CC(=O)N(C1=O)O
115.09
115.09
Liquid or Solid
98.0
93.0°C to 95.0°C
0.5% max. (60°C, 2 hrs, 1mm Hg)
98+%
MFCD00005516
01,487; 09,246
n-hydroxysuccinimide, hosu, 1-hydroxysuccinimide, 2,5-pyrrolidinedione, 1-hydroxy, 1-hydroxy-2,5-pyrrolidinedione, succinimide, n-hydroxy, n-hydroxysuccinimde, n-hydroxysuccinimid, unii-mje3791m4t, ccris 2604
NQTADLQHYWFPDB-UHFFFAOYSA-N
1-hydroxypyrrolidine-2,5-dione
80170
98+%
N-Hydroxysuccinimide, 98+%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 228-001-3

missing translation for 'tsca' : TSCA

Recommended Storage : This product contains some hydroxysuccinimide:ammonia adduct, which is virtually insoluble in acetone. The product has a tendency to caking.

Product Identifier
  • N-Hydroxysuccinimide

Signal Word
  • Warning

Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only