missing translation for 'onlineSavingsMsg'
Learn More

N-Iodosuccinimide, 98%

Product Code. 10185460 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10 g
50 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
10g
500g
50g
This item is not returnable. View return policy

Product Code. 10185460

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 516-12-1 | C4H4INO2 | 224.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 516-12-1
Molecular Formula C4H4INO2
Molecular Weight (g/mol) 224.99
MDL Number MFCD00005512
InChI Key LQZMLBORDGWNPD-UHFFFAOYSA-N
Synonym n-iodosuccinimide, succiniodimide, n-iodo succinimide, 2,5-pyrrolidinedione, 1-iodo, unii-3cos3x3n4p, nis, succinimide, n-iodo, 3cos3x3n4p, 1-iodo-pyrrolidine-2,5-dione, 1-iodoazolidine-2,5-dione
PubChem CID 120273
ChEBI CHEBI:53204
IUPAC Name 1-iodopyrrolidine-2,5-dione
SMILES IN1C(=O)CCC1=O

Specifications

Melting Point 197.0°C to 204.0°C
Color Brown to White-Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (Iodometry)
Packaging Glass bottle
Quantity 50 g
Merck Index 15,5088
Solubility Information Solubility in water: decomposes. Other solubilities: soluble in acetone,methanol,moderately soluble in dioxane,slightly soluble in methylene chloride and ethyl a,practically insoluble in carbon tetrachloride,eth
Formula Weight 224.99
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material N-Iodosuccinimide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.