missing translation for 'onlineSavingsMsg'
Learn More

N-Isopropylhydroxylamine hydrochloride, 97%, Thermo Scientific Chemicals

Product Code. 10679834
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50632-53-6
Molecular Formula C3H9NO·HCl
Molecular Weight (g/mol) 111.57
MDL Number MFCD00012599
InChI Key BYXUIKZQGOPKFR-UHFFFAOYSA-N
Synonym n-isopropylhydroxylamine hydrochloride, n-isopropyl hydroxylamine, n-isopropylhydroxylamine hcl, 2-propanamine, n-hydroxy-, hydrochloride, ccris 4317, i-propylhydroxylamine hydrochloride, isopropylhydroxylamine hydrochloride, n-hydroxypropan-2-amine hydrochloride, n-isopropyl hydroxylamine hydrochloride, n-isopropyl-hydroxylamine hydrochloride
PubChem CID 170873
IUPAC Name N-propan-2-ylhydroxylamine;hydrochloride
SMILES CC(C)NO.Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 68°C to 72°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 96% min. (Argentometry)
Packaging Glass bottle
Linear Formula (CH3)2CHNHOH·HCl
Quantity 1 g
Beilstein 04, 538
Formula Weight 111.57
Percent Purity 97%
Physical Form Adhering Crystalline Powder
Chemical Name or Material N-Isopropylhydroxylamine hydrochloride, 97%
Show More Show Less
compliance-icons
Product Identifier
  • N-Isopropylhydroxylamine hydrochloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'