missing translation for 'onlineSavingsMsg'
Learn More

N-Isopropylmethylamine, 98%, Thermo Scientific Chemicals

55.17 € - 217.00 €

Chemical Identifiers

CAS 4747-21-1
Molecular Formula C4H11N
Molecular Weight (g/mol) 73.14
InChI Key XHFGWHUWQXTGAT-UHFFFAOYSA-N
Synonym n-isopropylmethylamine, n-methylisopropylamine, methylisopropylamine, 2-propanamine, n-methyl, isopropylmethylamine, 2-methylaminopropane, methyl propan-2-yl amine, ethylamine, n,1-dimethyl, unii-j3ol90426g, n-methylisopropyylamine
PubChem CID 78485
IUPAC Name N-methylpropan-2-amine
SMILES CC(C)NC
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
11618459
View Documents
Thermo Scientific Acros
439650100
10 mL Glass bottle
55.17 €
10mL
Please to purchase this item. Need a web account? Register with us today!
11985181
View Documents
Thermo Scientific Acros
439650500
50 mL Glass bottle
217.00 €
50mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

4747-21-1
73.14
n-isopropylmethylamine, n-methylisopropylamine, methylisopropylamine, 2-propanamine, n-methyl, isopropylmethylamine, 2-methylaminopropane, methyl propan-2-yl amine, ethylamine, n,1-dimethyl, unii-j3ol90426g, n-methylisopropyylamine
N-methylpropan-2-amine
C4H11N
XHFGWHUWQXTGAT-UHFFFAOYSA-N
78485
CC(C)NC

Specifications

4747-21-1
100.0
0.7020g/mL
-32°C
98%
CH3NHCH(CH3)2
0.702
XHFGWHUWQXTGAT-UHFFFAOYSA-N
N-methylpropan-2-amine
78485
98%
N-Isopropylmethylamine
97.5
Colorless
50°C to 53°C
Authentic
C4H11N
1.3830 to 1.3850 (20°C, 589nm)
n-isopropylmethylamine, n-methylisopropylamine, methylisopropylamine, 2-propanamine, n-methyl, isopropylmethylamine, 2-methylaminopropane, methyl propan-2-yl amine, ethylamine, n,1-dimethyl, unii-j3ol90426g, n-methylisopropyylamine
CC(C)NC
73.14
73.14
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.
Toxic if swallowed.
Toxic if inhaled.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 225-266-7

Product Identifier
  • N-Isopropylmethylamine

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 A

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H301-Toxic if swallowed.
  • H331-Toxic if inhaled.
  • H225-Highly flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only