Learn More
N-Methyl-1-propylamine, 97%, Thermo Scientific Chemicals
27.54 € - 268.34 €
Chemical Identifiers
CAS | 627-35-0 |
---|---|
Molecular Formula | C4H11N |
Molecular Weight (g/mol) | 73.139 |
MDL Number | MFCD00009361 |
InChI Key | GVWISOJSERXQBM-UHFFFAOYSA-N |
Synonym | n-methylpropylamine, n-methyl-n-propylamine, methyl propyl amine, 1-propanamine, n-methyl, methyl-n-propylamine, methylpropylamine, unii-vw1h32h1qm, n-propylmethylamine, n-methyl-1-propylamine, vw1h32h1qm |
PubChem CID | 12315 |
IUPAC Name | N-methylpropan-1-amine |
SMILES | CCCNC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11468726
|
Thermo Scientific Alfa Aesar
A16903.03 |
1 g |
27.54 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11478726
|
Thermo Scientific Alfa Aesar
A16903.06 |
5 g |
73.87 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11488726
|
Thermo Scientific Alfa Aesar
A16903.14 |
25 g |
268.34 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
627-35-0 | |
73.139 | |
GVWISOJSERXQBM-UHFFFAOYSA-N | |
12315 | |
CCCNC |
C4H11N | |
MFCD00009361 | |
n-methylpropylamine, n-methyl-n-propylamine, methyl propyl amine, 1-propanamine, n-methyl, methyl-n-propylamine, methylpropylamine, unii-vw1h32h1qm, n-propylmethylamine, n-methyl-1-propylamine, vw1h32h1qm | |
N-methylpropan-1-amine |
Specifications
627-35-0 | |
62°C to 64°C | |
Amine-like | |
MFCD00009361 | |
n-methylpropylamine, n-methyl-n-propylamine, methyl propyl amine, 1-propanamine, n-methyl, methyl-n-propylamine, methylpropylamine, unii-vw1h32h1qm, n-propylmethylamine, n-methyl-1-propylamine, vw1h32h1qm | |
CCCNC | |
73.139 | |
73.14 | |
N-Methyl-1-propylamine |
0.713 | |
−1.1°C (30°F) | |
C4H11N | |
UN2733 | |
GVWISOJSERXQBM-UHFFFAOYSA-N | |
N-methylpropan-1-amine | |
12315 | |
97% |
Safety and Handling
GHS H Statement
H225-H314-H318
Highly flammable liquid and vapour.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H225-H302+H312+H332-H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.